Single crystal substrates
Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
A single crystal is a material in which the crystal lattice of the entire sample is continuous and unbroken to the edges of the sample, with no grain boundaries.
The absence of the defects associated with grain boundaries can give single crystals unique properties, particularly mechanical, optical and electrical. These properties, in addition to making them precious in some gems, are industrially used in technological applications, especially in optics and electronics.
Documents
Specifications
- Description:Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation
- Cat. No.:38509.KU
- Molecular formula:ZrO2 + 9.5% Y2O3
- MDL No.:MFCD01868885



