Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Single crystal substrates

Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation
Catalog # 38509.KT
Single crystal substrates
Catalog # 38509.KT
Single crystal substrates
Single crystal substrates

Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation

Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

A single crystal is a material in which the crystal lattice of the entire sample is continuous and unbroken to the edges of the sample, with no grain boundaries.

The absence of the defects associated with grain boundaries can give single crystals unique properties, particularly mechanical, optical and electrical. These properties, in addition to making them precious in some gems, are industrially used in technological applications, especially in optics and electronics.

Specifications
  • Description:
    Zirconium oxide/yttrium oxide substrate, 10×10×1 mm, polished one side, 100 orientation
  • Cat. No.:
    38509.KT
  • Molecular formula:
    ZrO2 + 9.5% Y2O3
  • MDL No.:
    MFCD01868885