Order Entry
Germany
Orders LinkContactUsLinkComponent
Ceramic plates, rectangular or square, AL25

Ceramic plates, rectangular or square, AL25

10 mm, 50 mm, 105 mm
Catalog # 33159.KF
Ceramic plates, rectangular or square, AL25
Catalog # 33159.KF
Ceramic plates, rectangular or square, AL25

10 mm, 50 mm, 105 mm

Price
175,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Pure Al₂O₃ Aluminium oxide, AL25.

Very porous, good thermal insulation, highest resistance to thermal shock of all the Al₂O₃ materials

Specifications
  • Width:
    50 mm
  • Length:
    105 mm
  • Seq:
    01
  • Thickness:
    10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.