Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Cyclohexane ≥99% for HPLC

Glass bottle, 1 L
Catalog # 22919.K2
Flammable
Health
Irritant
Environment
Cyclohexane ≥99% for HPLC
Catalog # 22919.K2
CAS Number:  110-82-7
Cyclohexane ≥99% for HPLC
Cyclohexane ≥99% for HPLC

Glass bottle, 1 L

CAS Number:  110-82-7
Price
81,50 €
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 99% min CAS No.: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -18°(-1°F) Physical Form: Liquid Density: 0.779 Refractive Index: 1.4260 Appearance: Colourless Odour: Pungent Melting Point: 6.5° Boiling Point: 80-81° Beilstein Reference: 1900225 Merck Reference: 14,2723 UN No.: 1145 MDL No.: MFCD00003814

Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:14,2723
  • Boiling Pt:80…81 °C
  • Melting Pt:6.5 °C
  • Density:0.779
  • Flash Pt:–18 °C (–1 °F)
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications