Graphite electrodes, counter
Graphite electrode, counter-spherical tip, 6,15×38,10 mm
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Graphite electrodes are used as homogeneous catalysts.
- AGKSP grade
- Ultra F purity
- Light sensitive and hygroscopic
- Incompatible with oxidising agents
- Applications include piston rings, thrust bearings, journal bearings and vanes
These are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.
Specifications
- Ø×L:6,15×38,10 mm
- Description:Graphite electrode, counter-spherical tip
- Seq:03