Titanium(III) fluoride
Plastic bottle for solids, 50 g
CAS No.: 13470-08-1 Molecular Formula: TiF3 Formula Weight: 104.90 Physical Form: Powder Density: 3.400 Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00042544
: Titanium trifluoride
- :13470-08-1
- :F₃Ti
- :MFCD00042544
- :104.88 g/mol
- :3.400
- :Ambient
- :236-732-4
- :3260
- :8,II
- :Density: 2.98 g/cm³ (20 °C)
- Pack type:Plastic bottle for solids
- Pk:50 g