Titanium(III) fluoride
Plastic bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 13470-08-1 Molecular Formula: TiF3 Formula Weight: 104.90 Physical Form: Powder Density: 3.400 Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00042544
Synonyms: Titanium trifluoride
- CAS Number:13470-08-1
- Formula:F₃Ti
- MDL Number:MFCD00042544
- Molecular Weight:104.88 g/mol
- Density:3.400
- Storage Temperature:Ambient
- EINECS:236-732-4
- UN:3260
- ADR:8,II
- :Density: 2.98 g/cm³ (20 °C)
Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:10 g



