Order Entry
Germany
Orders LinkContactUsLinkComponent
Vented caps for Falcon® flasks

Vented caps for Falcon® flasks

Vented caps for 25 cm² flasks
Catalog # 734-1066
   Low Manufacturing Impact
Supplier:   Corning
Vented caps for Falcon® flasks
Catalog # 734-1066
Supplier:  Corning
   Low Manufacturing Impact
Vented caps for Falcon® flasks
Vented caps for Falcon® flasks

Vented caps for 25 cm² flasks

Price
125,14 €
Log in for availability
1 * 100 ST
Product Details & Documents

PE with integral hydophobic 0,2 µm microporous membrane filter vent.

Vented caps minimise contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.

Specifications
  • Description:
    Vented caps for 25 cm² flasks
  • Cat. No.:
    734-1066
  • Environmental certification:
    ISO 14001 - Bedford
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.