Orders LinkContactUsLinkComponent
Urinary system model
Urinary system model
Prostate Model, 1/2 natural size
Catalog # BINH1000319
Marketsource
Supplier:   3B Scientific
Urinary system model
Catalog # BINH1000319
Supplier:  3B Scientific
Marketsource
Urinary system model
Urinary system model
Prostate Model, 1/2 natural size
Price
163,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Urinary system all-in-one model on baseboard showing:
  • Structures of retroperitoneal cavity
  • Large and small pelvis with bones and muscles
  • Inferior vena cava
  • Aorta with its branches including iliacal vessels
  • Upper urinary tract
  • Rectum
  • Kidney with adrenal gland
The prostate model provides a cross section of the male genital organs showing a healthy prostate with bladder, urethra, testicle, symphysis and rectum.
Specifications
  • Description:
    Prostate Model, 1/2 natural size