Rotary evaporating flasks
3000 ml, 29/32 NS
Catalog # 531-9127
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.
Specifications
- Socket size:29/32 NS
- Capacity:3000 ml
- Seq:11