Pestles
35 mm, 145 mm, 410-0083/410-0084
Catalog # 410-0086
Supplier: VITLAB
Jump to:
Product Details & Documents
Melamine-formaldehyde, white.
Specifications
- For:410-0083/410-0084
- Head Ø:35 mm
- Length:145 mm
- Cat. No.:410-0086
Melamine-formaldehyde, white.