Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Support rods, polished

12 mm, 450 mm, M 10
Catalog # RSGA240.204
Supplier:   RSG
Support rods, polished
Catalog # RSGA240.204
Supplier:  RSG
Support rods, polished
Support rods, polished

12 mm, 450 mm, M 10

Price
33,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polished rods made from stainless steel 18/10. Available with or without thread.

Specifications
  • Ø:
    12 mm
  • Thread:
    M 10
  • Length:
    450 mm
  • Seq:
    01