Orders LinkContactUsLinkComponent
Accessories
Accessories
pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membrane
Catalog # 18-1111-26
Supplier:   Cytiva
Accessories
Catalog # 18-1111-26
Supplier:  Cytiva
Accessories
Accessories
pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membrane
Price
1 470,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
    pH membrane
  • Cat. No.:
    18-1111-26
  • W×D×H:
    260×370×100 mm