Hoffmann pinch cocks
60 mm
Artikelnummer 229-0348
Hersteller: USBECK Laborgeräte
Jump to:
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
- Hinged
- Very good thermal resistance
- The standard model has a knurled screw and the heavy duty model has a butterfly screw
- Easy cleaning
Ordering information: The values in the band width column refer to the clearance between the pinch cock legs.
Specifications
- Bandwidth:60 mm