Quartz glass capsule in application temperatures of up to 1050°C (maximum 1600°C), 25 ml volume, circular, low form, translucent, h igh chemical resistance and purity. Diameter 54mm, height 15mm. 1 * 1 items
Catalog # TSLA73017
Supplier: TECHNICAL QUARTZ SOLUTIONS