Cis-reporting systems, PathDetect
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
- Identify pathway-specific transcription activation
- Easier and faster than blotting or gel-shift assays
- Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
: For research use only. Not for use in diagnostic procedures.
- Pk:1
- Description:PathDetect pCIS-CK plasmid
- Details:Negative control plasmid, recommended for use with all cis-reporter plasmids
- Contents:—