Orders LinkContactUsLinkComponent
Cis-reporting systems, PathDetect
PathDetect pLuc-MCS plasmid, PathDetect pLuc-MCS Multiple Cloning Site Vector, —, 50 µG
Catalog # STRA219087
Marketsource
Supplier:   STRATAGENE
Cis-reporting systems, PathDetect
Catalog # STRA219087
Supplier:  STRATAGENE
Marketsource
CAS Number:  
Cis-reporting systems, PathDetect
Cis-reporting systems, PathDetect
PathDetect pLuc-MCS plasmid, PathDetect pLuc-MCS Multiple Cloning Site Vector, —, 50 µG
CAS Number:  
Price
843,00 €
Log in for availability
1 * 50 µG
Product Details & Documents
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
 : For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    50 µG
  • Description:
    PathDetect pLuc-MCS plasmid
  • Details:
    PathDetect pLuc-MCS Multiple Cloning Site Vector
  • Contents:
Specifications