Cis-reporting systems, PathDetect
PathDetect p53 Cis-reporting System, System Includes: p53-Luc Cis-reporter plasmid and pFC-p53 control plasmid, p53-Luc plasmid (1 μg/μl) and pFC-p53 positive control plasmid (25 ng/μl), 1
Artikelnummer STRA219083
Marketsource
Hersteller: STRATAGENE
Cis-reporting systems, PathDetect
Artikelnummer STRA219083
Hersteller: STRATAGENE
Marketsource
CAS-Nummer:
Jump to:
Product Details & Documents
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
- Identify pathway-specific transcription activation
- Easier and faster than blotting or gel-shift assays
- Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
- Pk:1
- Description:PathDetect p53 Cis-reporting System
- Details:System Includes: p53-Luc Cis-reporter plasmid and pFC-p53 control plasmid
- Contents:p53-Luc plasmid (1 μg/μl) and pFC-p53 positive control plasmid (25 ng/μl)