Orders LinkContactUsLinkComponent
Trans-reporting systems, PathDetect
Trans-reporting systems, PathDetect
PathDetect Elk1 Trans-reporting system, 50 reactions, MAPK (mitogen-activated protein kinase) Pathway System Includes: pFR-Luc, pFA2-Elk1, pFC2-dbd and pFC-MEK1 plasmids, 1
Catalog # STRA219005
Marketsource
Supplier:   STRATAGENE
Trans-reporting systems, PathDetect
Catalog # STRA219005
Supplier:  STRATAGENE
Marketsource
CAS Number:  
Trans-reporting systems, PathDetect
Trans-reporting systems, PathDetect
PathDetect Elk1 Trans-reporting system, 50 reactions, MAPK (mitogen-activated protein kinase) Pathway System Includes: pFR-Luc, pFA2-Elk1, pFC2-dbd and pFC-MEK1 plasmids, 1
CAS Number:  
Price
1 480,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PathDetect trans-reporting systems allow you to determine if the protein or compound of interest directly or indirectly activates one of six transcription factors: c-Jun, Elk1, CREB, c-Fos, ATF2 or CHOP.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
Easily investigate whethe the protein or compound influences the activation of any of the six transcription factors offered and etermine the direct or indirect involvement of new gene products, growth factors, and drug candidates in specific pathways.
Delivery information: Contains of a reporter plasmid, a trans-activator plasmid and both positive and negative control plasmids.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    1
  • Description:
    PathDetect Elk1 Trans-reporting system
  • Size:
    50 reactions
  • Details:
    MAPK (mitogen-activated protein kinase) Pathway System Includes: pFR-Luc, pFA2-Elk1, pFC2-dbd and pFC-MEK1 plasmids