Order Entry
Germany
Orders LinkContactUsLinkComponent
ABLE C competent cells

ABLE C competent cells

ABLE C competent cells, Reduces the number of common cloning vectors 4-fold, 5×0,2 ml
Catalog # STRA200171
Marketsource
Supplier:   STRATAGENE
ABLE C competent cells
Catalog # STRA200171
Supplier:  STRATAGENE
Marketsource
ABLE C competent cells

ABLE C competent cells, Reduces the number of common cloning vectors 4-fold, 5×0,2 ml

Price
570,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.

  • Reduces the copy number of ColE1-derived plasmids (such as pUC and pBluescript® plasmids) four-fold
  • Decreases level of cloned protein product to increase cell viability
  • Transformation efficiency of 5×10⁶ to 2×10⁷ transformants/µg
  • Also available in an electroporation-competent version

ABLE C Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE C strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.

The ABLE C Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE C strains.

ABLE K Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE K strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.

ABLE K Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE K strains.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications
  • Description:
    ABLE C competent cells
  • Seq:
    03
  • Pk Info:
    5×0,2 ml
  • Feature:
    Reduces the number of common cloning vectors 4-fold
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.