Orders LinkContactUsLinkComponent
ABLE C competent cells
ABLE C competent cells
ABLE electroporation-competent cell kit, Includes both ABLE C and K cells, Contains: 5×0.1 ml ABLE C cells, 5×0.1 ml ABLE K cells., 1 kit
Catalog # STRA200160
Marketsource
Supplier:   STRATAGENE
ABLE C competent cells
Catalog # STRA200160
Supplier:  STRATAGENE
Marketsource
ABLE C competent cells
ABLE electroporation-competent cell kit, Includes both ABLE C and K cells, Contains: 5×0.1 ml ABLE C cells, 5×0.1 ml ABLE K cells., 1 kit
Price
923,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.
  • Reduces the copy number of ColE1-derived plasmids (such as pUC and pBluescript® plasmids) four-fold
  • Decreases level of cloned protein product to increase cell viability
  • Transformation efficiency of 5×10⁶ to 2×10⁷ transformants/µg
  • Also available in an electroporation-competent version
ABLE C Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE C strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
The ABLE C Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE C strains.
ABLE K Competent Cells for toxic clones enhance the probability of cloning DNA encoding toxic proteins. By reducing the copy number of ColE1-derived plasmids, the ABLE K strain decreases the levels of cloned gene products and enhances the probability of propagating a toxic clone.
ABLE K Competent Cells provide a viable alternative to commonly used cloning vectors with high copy numbers. Positive clones observed on initial screening of lambda libraries can be excised or re-cloned into any convenient vector and can be introduced into ABLE K strains.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    ABLE electroporation-competent cell kit
  • Seq:
    05
  • Pk Info:
    1 kit
  • Feature:
    Includes both ABLE C and K cells, Contains: 5×0.1 ml ABLE C cells, 5×0.1 ml ABLE K cells.