Lids for Nunc™ microplates
Lid, PS, with cut-off corners and evaporation barrier, for use with 384-well plates, Clear, 7,9 mm, Non sterile
Catalog # 732-2733
Supplier: Thermo Fisher Scientific
Lids for Nunc™ microplates
Catalog # 732-2733
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
- Sterility:Non sterile
- Colour:Clear
- Description:Lid, PS, with cut-off corners and evaporation barrier, for use with 384-well plates
- Height:7,9 mm
- Seq:08
- Cat. No.:732-2733
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact