Order Entry
Germany
Orders LinkContactUsLinkComponent
Extraction thimbles

Extraction thimbles

22×80 mm, 1,5 mm
Catalog # 533-0000
Extraction thimbles
Catalog # 533-0000
Extraction thimbles
Extraction thimbles

22×80 mm, 1,5 mm

Price
110,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.

Specifications
  • Int.Ø×H:
    22×80 mm
  • Seq:
    03
  • Thickness:
    1,5 mm