Order Entry
Germany
Orders LinkContactUsLinkComponent
Application specific filter papers, grade 287 ½, Whatman™

Application specific filter papers, grade 287 ½, Whatman™

287 ½, 150 mm
Catalog # 512-0581
Application specific filter papers, grade 287 ½, Whatman™
Catalog # 512-0581
Application specific filter papers, grade 287 ½, Whatman™
Application specific filter papers, grade 287 ½, Whatman™

287 ½, 150 mm

Price
69,40 €
Log in for availability
1 * 50 ST
Product Details & Documents

Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.

Specifications
  • Grade:
    287 ½
  • Seq:
    02
  • Size:
    150 mm
  • Thickness:
    360 µm
  • Cat. No.:
    512-0581
  • Filtration speed:
    330 s (Herzberg)
  • Weight:
    161 g/m²