ContactUsLinkComponent
FRACTOGEL EMD SO3- (S) STRONG CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (20-40 MYM) This product cannot be returne d after delivery 1 * 500 ml
Catalog # 1.16890.0500
Supplier:  Merck
FRACTOGEL EMD SO3- (S) STRONG CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (20-40 MYM) This product cannot be returne d after delivery 1 * 500 ml
Catalog # 1.16890.0500
Supplier:  Merck