ContactUsLinkComponent
FRACTOGEL EMD COO- (M) WEAK CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 5 l
Catalog # 1.16886.5000
Supplier:  Merck
FRACTOGEL EMD COO- (M) WEAK CATION EXCHANGER, SUSPENSION IN 20% ETHANOL AND 150 MM NACL (40-90 MYM) This product cannot be returned after delivery 1 * 5 l
Catalog # 1.16886.5000
Supplier:  Merck