Order Entry
Germany
Orders LinkContactUsLinkComponent
Standards, Spectroquant® CombiCheck, Supelco®

Standards, Spectroquant® CombiCheck, Supelco®

Spectroquant® CombiCheck 60, 125 ±13 mg/L Cl, 250 ±25 mg/L COD, 1 SET
Catalog # 1.14696.0001
Supplier:   Merck
Standards, Spectroquant® CombiCheck, Supelco®
Catalog # 1.14696.0001
Supplier:  Merck
CAS Number:  
Standards, Spectroquant® CombiCheck, Supelco®
Standards, Spectroquant® CombiCheck, Supelco®

Spectroquant® CombiCheck 60, 125 ±13 mg/L Cl, 250 ±25 mg/L COD, 1 SET

CAS Number:  
Price
161,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

Spectroquant® CombiCheck solutions are ready to use multi-parameter standards for the range of applications of Spectroquant® test system. Each pack contains a standard solution and a spiking solution. Both solutions can be used for quality assurance as they are, without dilution.

The standard solution can be used to confirm the accuracy of the results from the Spectroquant® system as a whole (photometer, reagents, analytical procedure, implementation). The spiking solution can be used to test for sample-dependent effects by measuring the recovery and confirms the suitability of the sample preparation technique.

The maximum number of determinations when using the standard solution depends upon the test system used.

Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    Spectroquant® CombiCheck 60
  • Range:
    125 ±13 mg/L Cl, 250 ±25 mg/L COD
  • Cat. No.:
    1.14696.0001
  • M-Clarity quality level:
    MQ100
  • Sulphate:
    -
  • Ammonium:
    -
  • Chloride:
    -
  • Nitrate:
    -
  • Phosphate:
    -
  • Model:
    Spectroquant® CombiCheck 60
  • COD:
    250 ±25 mg/L COD
  • Aluminium:
    -
  • Nickel:
    -
  • Manganese:
    -

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.