Crack sets for sample preparation, Spectroquant®, Supelco®
Catalog # 1.14688.0001
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Complete sets for digestion in a thermoreactor. The subsequent determination can be carried out in Spectroquant® test cells or reagent test sets. Crack Sets are easy to use and make sample preparation straightforward. For the preparation of dirty, coloured or samples with high levels of suspended material prior to using the appropriate Spectroquant® kit.
- Contains all reagents required for selected tests
- Easy to use - with step-by-step instructions
Documents
ProductPage_114687 ProductPage_114688 ProductPage_114963
Product specification
Specifications
Specifications
- Pk:25 Tests
- Description:Crack Set 10C Spectroquant®
- Cat. No.:1.14688.0001
- M-Clarity quality level:MQ100
- Parameters:Pb, Cd, Cr, Fe, Cu, Ni, Zn and P(total)
- Contents:Digestion reagent in round cells, acid, neutralising agent for pH adjustment.
- No. of digestions:25
- Model:Crack Set 10C
- Method:Peroxodisulfate/acidic



