Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer tablets according to Weise, Sigma-Aldrich® | Avantor

Buffer tablets according to Weise, Sigma-Aldrich®

6,8, Phosphate, 100 litres for 100 tablets, 100 Tab.
Catalog # 1.11374.0100
Supplier:   Merck
Buffer tablets according to Weise, Sigma-Aldrich®
Catalog # 1.11374.0100
Supplier:  Merck
CAS Number:  
Buffer tablets according to Weise, Sigma-Aldrich®
Buffer tablets according to Weise, Sigma-Aldrich®

6,8, Phosphate, 100 litres for 100 tablets, 100 Tab.

Wechseln zu:
CAS Number:  
Price
111,00 €
Anmelden für die Verfügbarkeit
1 * 100 Tablet
Product Details & Documents

Ideal for preparing buffer solution according to WEISE for staining of blood smears.

Prepared buffer solutions are used in hematology, for the dilution of staining solutions and for the rinsing steps in the staining process. Each tablet is sufficient for one litre of buffer solution, each produced solution can be stable for up to 4 weeks.

Spezifikationen:

Spezifikationen:

  • pH buffer type:
    Phosphate
  • Pk:
    100 Tab.
  • Final volume:
    100 litres for 100 tablets
  • pH:
    6,8

Spezifikationen: