Orders LinkContactUsLinkComponent
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
Catalog # 212-0006
Cryostorage boxes, Cryostore™
Catalog # 212-0006
Supplier:  
Cryostorage boxes, Cryostore™
Cryostorage boxes, Cryostore™
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Cryobox 81 places for External threaded vials, made from polu carbonate with drin slots, capacity up to 1-2ml, Blue, 1 * 5 items Catalog #479-2395
Product 479-2395 is similar to 212-0006 but manufacturer and specifications may vary.
Cryo Boxes with Grid, colored polycarbonate (PC) green. Format W 132 x D 132 mm x H 52 mm. 81 places numerically coded. 1 * 5 items Catalog #RATI5150002
Product RATI5150002 is similar to 212-0006 but manufacturer and specifications may vary.
Product Details & Documents
PC, coloured.
  • Writable surfaces
  • Available with four colours
  • Printed with squared numbers
  • Transparent cover allows to see the content of the box
These durable cryogenic storage boxes are designed to be used at temperatures between −196 °C and +121 °C. It can accommodate around 42×T310-10A Cryovials®.
Documents