Order Entry
Germany
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Histosette® II, M492

Tissue processing/embedding cassettes, Histosette® II, M492

White
Catalog # 720-0909
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Catalog # 720-0909
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Tissue processing/embedding cassettes, Histosette® II, M492

White

Price
336,00 €
Log in for availability
1 * 1.500 ST
Product Details & Documents

Acetal polymer.

  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
Specifications
  • Colour:
    White