Orders LinkContactUsLinkComponent
GC capillary column tools
Replacement PTFE 1/16" sealing ring
  558-0190
GC capillary column tools
  558-0190
GC capillary column tools
Replacement PTFE 1/16" sealing ring
53,20 €
1 * 20 ST
The capillary cutting tool is used for burr free cutting of fused silica tubing.
  • Reduces likelihood of blockages
  • Ensures square ends for 'Press fit' style connectors
  • Clean entry for on-column injection
  • Description:
    Replacement PTFE 1/16" sealing ring
  • Seq:
    02