Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Sodium azide

Plastic bottle, 250 g
Catalog # 1.06688.0250
Toxic
Health
Environment
Supplier:   Merck
Sodium azide
Catalog # 1.06688.0250
Supplier:  Merck
CAS Number:  26628-22-8
Sodium azide

Plastic bottle, 250 g

CAS Number:  26628-22-8
Price
82,60 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:13,08653
  • Melting Pt:275 °C
  • Density:1.85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.