ContactUsLinkComponent
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) TAPERING TO 0.47 MM OD (26 GAUGE) AT THE TIP 42 MM LONG W/ CONE POINT STYLE 1 * 1 items
Catalog # 549-0393
SGE SYRINGE 10µL REMOVABLE NEEDLE FOR AGILENT 7673/7683 AUTOSAMPLER NEEDLE IS STAINLESS STEEL 0.63 MM OD (23 GAUGE) TAPERING TO 0.47 MM OD (26 GAUGE) AT THE TIP 42 MM LONG W/ CONE POINT STYLE 1 * 1 items
Catalog # 549-0393
Frequently Bought Together