ContactUsLinkComponent
SGE SYRINGE 10µL FIXED NEEDLE THAT IS 0.47 MM OD (26 GAUGE) 50 MM LONG W/ BEVEL POINT STYLE WITH REPEATING ADAPTER 1 * 1 items
Catalog # 549-0436
SGE SYRINGE 10µL FIXED NEEDLE THAT IS 0.47 MM OD (26 GAUGE) 50 MM LONG W/ BEVEL POINT STYLE WITH REPEATING ADAPTER 1 * 1 items
Catalog # 549-0436
Frequently Bought Together