Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Creatinine, Millipore®

Plastic bottle, 50 g
Catalog # 1.05206.0050
Supplier:   Merck
Creatinine, Millipore®
Catalog # 1.05206.0050
Supplier:  Merck
CAS Number:  60-27-5
Creatinine, Millipore®

Plastic bottle, 50 g

CAS Number:  60-27-5
Price
79,90 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-466-7
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g

Specifications