Tween® 80 (Polysorbate)
Glass bottle for solids, 500 g
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:∼100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1.062 g/cm³ (20 °C)
- Flash Pt:>149 °C
- Storage Temperature:Ambient
- EINECS:500-019-9
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



