Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Plastic bottle for solids, 500 g
Catalog # SERA37470.01
Marketsource
Supplier:   Serva
Tween® 20 (Polysorbate)
Catalog # SERA37470.01
Supplier:  Serva
Marketsource
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Plastic bottle for solids, 500 g

CAS Number:  9005-64-5
Price
51,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.