Order Entry
Germany
Orders LinkContactUsLinkComponent
Paraformaldehyde

Paraformaldehyde

Glass bottle for solids, 500 g
Catalog # SERA31628.02
Flammable
Health
Irritant
Marketsource
Supplier:   Serva
Paraformaldehyde
Catalog # SERA31628.02
Supplier:  Serva
Marketsource
CAS Number:  30525-89-4
Paraformaldehyde

Glass bottle for solids, 500 g

CAS Number:  30525-89-4
Price
64,30 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:123 °C
  • Density:1.46 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.