Order Entry
Germany
Orders LinkContactUsLinkComponent
Mitomycin C

Mitomycin C

Glass bottle for solids, 20 mg
Catalog # SERA29805.03
Toxic
Health
Marketsource
Supplier:   Serva
Mitomycin C
Catalog # SERA29805.03
Supplier:  Serva
Marketsource
CAS Number:  50-07-7
Mitomycin C

Glass bottle for solids, 20 mg

CAS Number:  50-07-7
Price
Inquire For Price
Log in for availability
1 * 20 mg
Product Details & Documents
Synonyms:  Mitomycin, Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • MDL Number:MFCD00078109
  • Molecular Weight:334.33 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-008-6
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    20 mg

Specifications