Mitomycin C
Glass bottle for solids, 10 mg
Catalog # SERA29805.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Mitomycin, Ametycin
- CAS Number:50-07-7
- Formula:C₁₅H₁₈N₄O₅
- MDL Number:MFCD00078109
- Molecular Weight:334.33 g/mol
- Storage Temperature:Refrigerator
- EINECS:200-008-6
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg



