Order Entry
Germany
Orders LinkContactUsLinkComponent
Mitomycin C

Mitomycin C

Glass bottle for solids, 2 mg
Catalog # SERA29805.01
Toxic
Health
Marketsource
Supplier:   Serva
Mitomycin C
Catalog # SERA29805.01
Supplier:  Serva
Marketsource
CAS Number:  50-07-7
Mitomycin C

Glass bottle for solids, 2 mg

CAS Number:  50-07-7
Price
145,00 €
Log in for availability
1 * 2 mg
Product Details & Documents
Synonyms:  Mitomycin, Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • MDL Number:MFCD00078109
  • Molecular Weight:334.33 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:200-008-6
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.