Orders LinkContactUsLinkComponent
D-(+)-Mannose
Glass bottle for solids, 50 g
  SERA28460.02
Marketsource
 :   Serva
D-(+)-Mannose
  SERA28460.02
 :  Serva
Marketsource
 :  3458-28-4
D-(+)-Mannose
Glass bottle for solids, 50 g
 :  3458-28-4
95,60 €
1 * 50 g
 :  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  •  :3458-28-4
  •  :C₆H₁₂O₆
  •  :MFCD00064122
  •  :180.16 g/mol
  •  :13,05772
  •  :376 °C (1013 hPa)
  •  :133 °C
  •  :1.539 g/cm³ (20 °C)
  •  :Ambient
  •  :222-392-4
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g