Order Entry
Germany
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Glass bottle for solids, 50 mg
Catalog # SERA28073.03
Marketsource
Supplier:   Serva
D(-)-Luciferin
Catalog # SERA28073.03
Supplier:  Serva
Marketsource
CAS Number:  2591-17-5
D(-)-Luciferin

Glass bottle for solids, 50 mg

CAS Number:  2591-17-5
Price
Inquire For Price
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • EINECS:219-981-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications