Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Imidazole

Glass bottle for solids, 25 g
Catalog # SERA26080.02
Corrosive
Health
Irritant
Marketsource
Supplier:   Serva
Imidazole
Catalog # SERA26080.02
Supplier:  Serva
Marketsource
CAS Number:  288-32-4
Imidazole
Imidazole

Glass bottle for solids, 25 g

CAS Number:  288-32-4
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:13,04935
  • Boiling Pt:255 °C (1013 hPa)
  • Melting Pt:89…90 °C
  • Density:1.18 g/cm³ (20 °C)
  • Flash Pt:135 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications