Orders LinkContactUsLinkComponent
Base materials
Base materials
Yeast extract, Servabacter, powder, Glass bottle for solids, 500 g
Catalog # SERA24540.02
Marketsource
Supplier:   Serva
Base materials
Catalog # SERA24540.02
Supplier:  Serva
Marketsource
CAS Number:  
Base materials
Yeast extract, Servabacter, powder, Glass bottle for solids, 500 g
CAS Number:  
Price
105,00 €
Log in for availability
1 * 500 g
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Description:
    Yeast extract, Servabacter, powder