Orders LinkContactUsLinkComponent
Wright's stain solution
Wright's stain solution
Plastic bottle, Preparation Capacity 5000 slides, 2,5 L
  1.01383.2500
 :   Merck
Wright's stain solution
  1.01383.2500
 :  Merck
 :  68988-92-1
Wright's stain solution
Plastic bottle, Preparation Capacity 5000 slides, 2,5 L
 :  68988-92-1
167,00 €
1 * 2,5 L
Used to stain blood, bone marrow smears, and clinical cytological specimens
The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.
 :  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  •  :68988-92-1
  •  :MFCD00082143
  •  :0.80 g/cm3 (20 °C)
  •  :9.7 °C
  •  :1230
  •  :3 (6.1),II
Documents
  • Suitability for microscopy (Blood smear):
    passes test
  • Erythrocytes:
    pink to brownish
  • Nuclei:
    blue-violet
  • Eosinophilic granules:
    red to red-brown
  • Neutrophilic granules:
    light violet
  • Lymphocyte cytoplasm:
    Blue
  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Pk Info:
    Preparation Capacity 5000 slides