Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Wright's stain solution

Plastic bottle, 500 mL
Catalog # 1.01383.0500
Flammable
Toxic
Health
Supplier:   Merck
Wright's stain solution
Catalog # 1.01383.0500
Supplier:  Merck
CAS Number:  68988-92-1
Wright's stain solution

Plastic bottle, 500 mL

CAS Number:  68988-92-1
Price
55,60 €
Log in for availability
1 * 500 mL
Product Details & Documents

Used to stain blood, bone marrow smears, and clinical cytological specimens

The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.

Synonyms:  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  • CAS Number:68988-92-1
  • MDL Number:MFCD00082143
  • Density:0.80 g/cm3 (20 °C)
  • Flash Pt:9.7 °C
  • UN:1230
  • ADR:3 (6.1),II
Specifications

Specification Test Results

  • Suitability for microscopy (Blood smear):
    passes test
  • Erythrocytes:
    pink to brownish
  • Nuclei:
    blue-violet
  • Eosinophilic granules:
    red to red-brown
  • Neutrophilic granules:
    light violet
  • Lymphocyte cytoplasm:
    Blue

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications