Order Entry
Germany
Orders LinkContactUsLinkComponent
Brij® 35

Brij® 35

Glass bottle for solids, 100 g
Catalog # SERA15230.01
Irritant
Marketsource
Supplier:   Serva
Brij® 35
Catalog # SERA15230.01
Supplier:  Serva
Marketsource
CAS Number:  9002-92-0
Brij® 35

Glass bottle for solids, 100 g

CAS Number:  9002-92-0
Price
34,70 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Polyoxyethylene(23)lauryl ether, Tricosaethylene glycol mono-n-dodecyl ether, Brij® L23
  • CAS Number:9002-92-0
  • Formula:C₅₈H₁₁₈O₂₄
  • MDL Number:MFCD00080891
  • Molecular Weight:1199.56 g/mol
  • Storage Temperature:Ambient
  • EINECS:500-002-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications