Ampicillin
Glass bottle for solids, 25 g
Catalog # SERA13397.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
- EINECS:200-709-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



